Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®

Supplier: EMD MILLIPORE # PCS CA EXCEP
Warning

1.10164.9025 1.10164.1000 1.10164.2500 1.10164.9950
CA1.10164.9025EA 1740.97 CAD
CA1.10164.9025 EM1.10164.1000 CA1.10164.2500 CA1.10164.9950
Hydrochloric acid 6 mol/l, EMPROVE® EXPERT, SAFC®
Hydrochloric acid

Manufactured from hydrochloric acid fuming 37% EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF and purified water acc. Ph. Eur., JP.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: HCl
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,

Order Now

Specification Test Results

Concentration (HCl) 5.7 - 6.3 mol/l
Identity Passes Test
Color ≤10 Hazen
Appearance Passes Test
Free Bromine or Free Chlorine Passes Test
Free Chlorine (Cl) ≤0.0001
Sulfate (SO₄) ≤0.0005%
Sulfite (SO₃) ≤0.001%
Heavy Metals (as Pb) ≤0.0001%
As (Arsenic) ≤0.00015%
Residual Solvents (ICH Q3C) Excluded by Production Process
Residue on Ignition (as SO₄) ≤0.008%
Evaporation Residue ≤0.01
Endotoxins ≤0.25 I.U./ml
Filtered with 0.2 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR