Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL JPC, USP, Ph. Eur., BP, FCC, E511, SAFC®

Supplier: MilliporeSigma

Synonyms: Magnesium dichloride

1.05832.1000 1.05832.5000 1.05832.5000 1.05832.9029 1.05832.9525
EM1.05832.1000EA 543.08 CAD
EM1.05832.1000 EM1.05832.5000 CA1.05832.5000 CA1.05832.9029 CA1.05832.9525
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), crystals, EMPROVE® ESSENTIAL JPC, USP, Ph. Eur., BP, FCC, E511, SAFC®
Magnesium chloride hexahydrate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.


Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117 Dec.
Density: 1.57 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay (Complexometric) 99.0 - 101.0%
Identity Passes Test
Appearance of Solution Passes Test
Insoluble Matter ≤0.005%
Acidity or Alkalinity Passes Test
pH-value (5%; Water) 5.0 - 7.0
Bromide (Br) ≤0.05%
Sulfate (SO₄) ≤0.005%
Heavy Metals (as Pb) ≤0.001%
Al (Aluminium) ≤0.0001%
As (Arsenic) ≤0.0002%
Ba (Barium) Passes Test
Ca (Calcium) ≤0.01%
Fe (Iron) ≤0.0005%
Hg (Mercury) ≤0.0001%
K (Potassium) ≤0.0500%
NH₄ (Ammonium) ≤0.005%
Pb (Lead) ≤0.0002%
Residual Solvents (Ph. Eur./USP/ICH) Excluded by Manufacturing Process
Water 51.0 - 55.0%
Endotoxins ≤3.0 I.U./g
Total Aerobic Microbial Count ≤100 CFU/g
Total Yeast and Mould Count ≤100 CFU/g
Corresponds to Ph. Eur., BP, USP, JPC, FCC, E511


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR