Bulk-Pak™ Serological Pipettes with Cotton Plugging Top, Bellco Glass
Supplier: Bellco Glass
1200-01001 1200-02010 1200-10010 1200-05010 1200-25010
CA12101-002CS 10935.4 CAD
CA12101-002 CA12101-008 CA12101-004 CA12101-006 CA12101-010
Bulk-Pak™ Serological Pipettes with Cotton Plugging Top, Bellco Glass
Pipettes Serological Pipettes
Constricted top for cotton plugging.
  • Type 1 Class B borosilicate glass
  • Tempered tip; to deliver, blow out
Plain top promotes rapid even flow with a calibrated tip. All Bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings.
Delivery information: Supplied without cotton.