VWR® Utility Support Uprights
Supplier: VWR
VWR®
USUSP13-SW USUM1500-SW USUHCM1800-VWR USUM1800-VWR USUSP11-VWR USUFPM1804-SW USUDIV-VWR USUSP14-VWR USUFPM1504-SW USUSP13-VWR USUM1200-VWR USUFPM1203-SW USUHCM1200-SW USUFPM1204-SW USUDIV-SW USUHCM1500-VWR USUM1200-SW USUSP14-SW USUFPM1805-VWR USUFPM1204-VWR USUHCM1500-SW USUHCM1800-SW USUFPM1804-VWR USUFPM1803-VWR USUM1800-SW USUFPM1505-SW USUFPM1205-VWR USUHCM1200-VWR USUFPM1503-SW USUSP11-SW USUFPM1205-SW USUFPM1503-VWR USUFPM1803-SW USUM1500-VWR USUFPM1505-VWR USUFPM1805-SW USUSP12-SW USUFPM1203-VWR USUFPM1504-VWR USUSP12-VWR
76308-940EA 117.71 CAD
76308-940 76551-078 76308-842 76308-864 76551-054 76551-154 76308-866 76551-196 76308-882 76308-884 76308-840 76308-928 76550-708 76308-868 76550-752 76550-510 76550-998 76550-494 76550-472 76550-596 76308-874 76551-024 76308-930 76551-142 76308-876 76308-870 76308-872 76308-838 76550-738 76308-938 76308-878 76308-934 76308-936 76550-600 76550-480 76308-880 76550-980 76550-420 76550-960 76550-564
VWR® Utility Support Uprights
Furniture Casework
USU Utility Support Uprights are the ideal solution to adding utility support above the work surface.
  • Powder coated steel
  • Attach directly to a top and allow for easy access to utilities without reducing the usable work surface area
  • Available in three width options, and can be equipped with optional shelves, horizontal channels, and dividers
  • Services can be connected via overhead service carriers, or from underneath the surface through the mounting bracket
These adjustable cable channels can be configured to suit a range of power, gas, and data needs, while also providing shelving storage above the worksurface. Cable channels hide cords and hoses, creating a clean work space.
Ordering information: Bookend brackets are required for attachment of shelves to USU system. Service fixtures, cables, hoses, etc. are not included with the USU. These are ordered separately. Work surfaces must be ordered according to specifications below. Please consider hole locations and dimensions of USU when configuring for your space. Work surfaces are larger than standard work surfaces in order to support width of USU system. USU will not attach to VWR® benches without a work surface with correct dimensions. Contact your Avantor representative for complete work surface specification files, required accessories, or for more information.
Horizontal utility channel includes a blank front panel and bottom cover. Additional panels available for front-facing utility support. Horizontal utility channel mounts to single bay USU system. Adding a middle divider to this channel creates a double bay system. Middle vertical divider is required for attachment of M30 and M36 shelves. Divider and shelves do not contain utility support. Utility support is housed in vertical USU uprights and horizontal channel only.
Delivery information: Single Bay Starter Kits include two vertical USU uprights, blank service panels with attachment hardware, fixed horizontal utility channel/shelf with blank front panel, and upper horizontal cable channel with cover (blank panels do not include cut-outs for connecting services; to add services, please select the panel according to services required).