VWR® Pierceable, Silicone Microplate Sealing Mats
Supplier: VWR International
76311-634 76311-636 76311-638 76311-640
76311-634CS 1293.07 CAD
76311-634 76311-636 76311-638 76311-640
VWR® Pierceable, Silicone Microplate Sealing Mats
Seals Microplate Seals
Suitable for sealing microplates in incubation and storage applications, silicone sealing mats provide sample protection while minimizing evaporation in a wide range of temperatures.
  • Pierceable for easy penetration; self-healing
  • Chemically resistant, research-grade silicone
  • Contains no adhesives or extractables to contaminate samples
  • Printed alpha numeric grid for easy reference
  • Eliminates cross-contamination between wells
  • Sample access via pipettor, syringe, or probe
  • Airtight closure helps minimize sample evaporation
  • Resists coring and tearing
Silicone microplate sealing mats are suitable for high-throughput applications and high-temperature storage. All mats are reusable; autoclave and follow with 10% bleach wash and ethanol rinse.
Certifications: Certified Rnase-, Dnase-, DNA- and Pyrogen-free.