KIMBLE® GLS 80® Media Bottles, Amber, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
14394-0250 14394-0500 14394-1000 14394-2000 14394-5000 14394-10L 14394-20L
76312-284CS 702.5 CAD
76312-284 76312-286 76312-288 76312-290 76312-292 76312-294 76312-296
KIMBLE® GLS 80® Media Bottles, Amber, Wide Mouth, with Blue Polypropylene Caps, DWK Life Sciences
Bottles Glass Bottles
Get the versatility, purity, and durability needed to achieve results for demanding laboratory storage, bioprocess, and production applications.
  • Blue, quick-release, linerless polypropylene closure and pour ring are rated up to 140 °C
  • Amber tinted bottles provide UV light protection up to 500 nm
  • Autoclavable
Ideal for storage and transport of high-value sample substances, with a wide-mouth design that provides ease of use for filling and dispensing of powders, viscous liquids, and slurries. Durable, fired on ceramic ink graduations and marking patch withstand the rigors of daily laboratory, bioprocess, and production use. KIMBLE® 3.3 borosilicate glass is low-extractable, USP Type 1, with a full temperature and chemical resistance range.