WHEATON® BlueMAG™ Polypropylene Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
18-4100 18-4101 18-4102 18-4103 18-4104 18-4105 09-4100 09-4101 09-4102
76313-686PK 141.85 CAD
76313-686 76313-688 76313-690 76313-692 76313-694 76313-696 76313-698 76313-700 76313-702 76313-704
WHEATON® BlueMAG™ Polypropylene Screw Caps, DWK Life Sciences
Closures Chromatography Vial Closures
Uniquely designed for automated GC, GC-MS, and LC-MS chromatography magnetic sample transport systems.
  • Available in 18 mm headspace septa and 9 mm screw caps
  • Crimped surface provides both a flat, squared surface for machine aoutomation
  • Cap are compatible with all major brands of autosamplers, X-Y-Z robotics used in GC-MS or LC-MS applications
Magnetic automation transport systems require robust performance and sometimes, even with careful attention to detail, some caps do not sit perfectly square to the vial axis. When this occurs, the magnetic systemMore Product Information