Wheaton® MicroLiter® ULTRAPure Septa Crimp Caps and Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
09-0038N 09-0039N 11-0038A 11-0039A
76045-380CS 75.28 CAD
76045-380 76045-382 76288-112 76288-114
Wheaton® MicroLiter® ULTRAPure Septa Crimp Caps and Screw Caps, DWK Life Sciences
Closures Chromatography Vial Closures
Caps are manufactured from the highest purity, ultra-low bleed silicone available in the market.
  • 9 mm natural polypropylene caps or 11 mm silver aluminum crimp caps
  • Available in non-slit and pre-slit formats
  • Ultra-low bleed blue/teal silicone Shore A is laminated to white PTFE film, 3 mm thick
  • Silver aluminum crimp caps are lacquer coated
Certified ULTRAPure septa is manufactured from the highest purity, ultra-low bleed silicone available in order to combat the leachable effects of silicone without trading off the ability to be pierced multiple times. The MicroLiter certified pure septa provides accurate and reproducible LC-MS and GC-MS results.