Potassium hydroxide 0.1 M KOH (0.1N) Volumetric, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Potash caustic, Caustic potash

35125-10L-VP 35125-1L
BJ35125-10L-VPEA 950.09 CAD
BJ35125-10L-VP BJ35125-1L
Potassium hydroxide 0.1 M KOH (0.1N) Volumetric, Fluka™
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
Merck Index: 12,07806

Order Now

Specification Test Results

Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR