n-Pentane ≥98.0%, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Pentane

NS312-20 NS312-20
CABJNS312-20EA 2707.91 CAD
CABJNS312-20 BJNS312-20
n-Pentane ≥98.0%, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
n-Pentane

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 7.8 %(V)
Melting Pt: –129.7 °C
Density: 0.626 g/cm³ (20 °C)
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
https://ca.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
GC Analysis (n-Pentane) 98 % min.
GC Analysis (total C5 isomers) 99.9 % min.
Refractive Index (20ºC) 1.3573 - 1.3579
Residue 1 mg/L max.
UV Absorbance @ 200 nm 0.6 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Cutoff 190 nm max.
Water by Karl Fischer Titration 0.01 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR