Iodine 0.05 M I2 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

35090-6X1L 35090-4X2.5L
BJ35090-1LCS 542.27 CAD
BJ35090-1L BJ35090-2.5L
Iodine 0.05 M I2 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Iodine
Formula: I₂
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR