EDTA disodium salt 0.01 M EDTA-Na2 volumetric solution, for complexometry, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

35322-1L 35322-500ML
BJ35322-1LEA 329.89 CAD
BJ35322-1L BJ35322-500ML
EDTA disodium salt 0.01 M EDTA-Na2 volumetric solution, for complexometry, Fluka™
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 231-791-2
Merck Index: 12,03556

Order Now

Specification Test Results

Factor 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR