Disodium oxalate 0.05 M (COO)2Na2 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

35240-1L 35240-20L
BJ35240-1LEA 79.65 CAD
BJ35240-1L BJ35240-20L
Disodium oxalate 0.05 M (COO)2Na2 volumetric solution, Fluka™
di-Sodium oxalate
Formula: Na₂C₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 231-791-2

Order Now

Specification Test Results

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR