Macro Cuvettes, FireflySci
Supplier: FIREFLYSCI, INC.
5G10 5UV10 61FLIR10 62G10 62UV10 62FLG10 62FLUV10 64UV10 552UV10X1 552UV10X2 552UV10X3 608FLUV3X3X11A 608FLUV3X3X11B 50UV5 50UV10 51HUV10 51VUV10 51VIR10 52G10X1 52G10X2 52G10X3 52G10X4 52G10X5 53UV10 53IR10 54UV10 54IR10 54FLUV10 54FLIR10 56UV10 56IR10 58UV10 58IR10 58FLUV10 59FLUV10 59FLIR10 59MUV10 59MIR10 60UV10 61UV0.5 61UV1 61UV2 61UV5 61UV10 61IR0.5 61IR1 61IR2 61IR5 61IR10 61FLUV10 64IR10 64FLUV10 64FLIR10 65FLUV10 509G10 509UV10 510UV 511UV 512UV 513MIR10 514MUV10 514MIR10 515AUV10 520MUV5 520MUV10 520MUV20 520MIR5 520MIR10 520MIR20 521G100 521UV100 522G10 522UV10 529G5 526UV0.25 552UV10X4 552UV10X5 601UV1A 601UV1B 601UV2A 601UV2B 601UV5A 601UV5B 601UV10A 601UV10B 602UV10A 602UV10B 603FLUV1X1X11B 603FLUV3X3X11B 603FLUV10X2X2B 603FLUV10X2X2C 603FL10X4X11B 605UV1A 605UV1B 605UV2A 605UV2B 606UV5A 606UV5B 606UV10A 606UV10B 607UV3X3X27 607UV3X3X43 607IR3X3X27 607IR3X3X43 608FLUV1X1X11A 608FLUV1X1X11B 6081.5X1.5X11A 6081.5X1.5X11B
76015-196EA 106.71 CAD
76015-196 76015-198 76015-766 76015-768 76015-770 76015-772 76015-774 76015-776 76015-778 76015-780 76015-782 76016-108 76016-110 76016-208 76016-210 76016-212 76016-216 76016-218 76016-220 76016-222 76016-224 76016-226 76016-228 76016-250 76016-252 76016-254 76016-256 76016-258 76016-260 76016-262 76016-264 76016-266 76016-268 76016-270 76016-272 76016-274 76016-276 76016-278 76016-280 76016-282 76016-284 76016-286 76016-288 76016-290 76016-292 76016-294 76016-296 76016-298 76016-300 76016-302 76016-304 76016-306 76016-308 76016-310 76016-316 76016-414 76016-416 76016-418 76016-420 76016-422 76016-426 76016-428 76016-430 76016-432 76016-434 76016-436 76016-438 76016-440 76016-442 76016-444 76016-446 76016-448 76016-450 76016-452 76016-456 76016-458 76016-460 76016-462 76016-464 76016-466 76016-468 76016-470 76016-472 76016-474 76016-476 76016-478 76016-480 76016-482 76016-484 76016-486 76016-488 76016-490 76016-492 76016-494 76016-496 76016-498 76016-500 76016-502 76016-504 76016-506 76016-508 76016-510 76016-512 76016-514 76016-516 76016-518 76016-520 76016-522 76016-524
Macro Cuvettes, FireflySci
Cuvettes Spectrophotometer Cuvettes
Firefly's black quartz micro fluorescence flow cell.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in optical glass, IR quartz and UV quartz
These cuvettes are a good fit for many researchers with custom builds and machines. Highly accurate results, only need to test sample once. Firefly cuvettes are made with the best quality quartz and glass material available. Has excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a vigorous inspection process before leaving the factory.
Caution: Cuvettes must be scrupulously clean. Optical surfaces should not be touched as oil smudges are difficult to remove. Purity of solvents used for sample and cleaning is important. Only distilled water or spectrophotometric grade solvents should be used to avoid contamination from impure solvents. As soon as possible after use, cuvettes should be rinsed and soaked in distilled water.