Macro Flow Through Cells with Detachable Tubes, FireflySci
Supplier: FIREFLYSCI, INC.
45UV10 45IR10
76016-142EA 1421.13 CAD
76016-142 76016-144
Macro Flow Through Cells with Detachable Tubes, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality macro cuvettes comes with spectrophotometer cells with macro capacities and are a great value.
  • Comes in several different materials: (UV quartz and IR quartz)
  • Available in 10 mm lightpath
  • Two windows polished
  • Tubes are made of quartz
These macro spectrophotometer flow through cell with detachable inlet/outlet quartz tubes. Enables a constant flow through the 2.8 ml capacity observation chamber. Highly accurate results, only need to test sample once. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a rigorous inspection process before leaving our factory. Fire-fused to ensure that cuvettes are robust and durable.