Macro Fluorescence Cuvettes with Screw Cap, FireflySci
Supplier: FIREFLYSCI, INC.
41G5 41G10 41G50 41G100 41FLG5 41FLG10 41FLG50 41FLG100
76016-050EA 511.16 CAD
76016-050 76016-052 76016-054 76016-056 76016-070 76016-072 76016-074 76016-076
Macro Fluorescence Cuvettes with Screw Cap, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality macro fluorescence cuvette with screw cap comes in several different materials and are a great value.
  • Available in pyrex glass and UV quartz
  • Suitable for anaerobic and dye laser work
  • Rated to handle up to 5 bar of pressure
These Type 41FL cuvette is our standard fluorescence screw cap cell and is a favorite among scientists who perform anaerobic work. The open SEPTA screw cap enables researchers to make sterile injections into the cuvette without compromising the experiment. Lightpaths of 5 to 100 mm available. Four windows and base optically polished. Features 13-425 screw cap threadings for maximum durability and versatility. Fire-fused to ensure that cuvettes are robust and durable.
Delivery information: Includes 1 SEPTA screw cap and 1 closed screw cap.