Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
Supplier: FIREFLYSCI, INC.
3FTG10 3FTUV10 3FTIR10
76015-178EA 559.75 CAD
76015-178 76015-180 76015-182
Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality macro open bottom fluorescence flow through are of a great value.
  • Comes in several different materials: (Optical Glass, UV quartz, IR quartz)
  • Available in 10 mm lightpath
  • Four windows optically polished
  • Both ends are open
Fire-fused to ensure that cuvettes are robust and durable. Highly accurate results, only need to test sample once. These cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes throughMore Product Information