Cuvettes with Graded Seal Tube, FireflySci
Supplier: FIREFLYSCI, INC.
37G1 37G2 37G5 37G10 37G20 37G50 37G100 37UV1 37UV2 37UV5 37UV10 37UV20 37UV50 37UV100 37IR1 37IR2 37IR5 37IR10 37IR20
76016-012EA 334.23 CAD
76016-012 76016-014 76016-016 76016-018 76016-020 76016-022 76016-024 76016-026 76016-028 76016-030 76016-032 76016-034 76016-036 76016-038 76016-040 76016-042 76016-044 76016-046 76016-048
Cuvettes with Graded Seal Tube, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality cuvettes with quartz-to-pyrex graded seal tubes and are a great value.
  • Comes in several different materials: (Optical Glass, UV quartz, IR quartz)
  • Lightpaths from 0.05 - 5mm available
  • Two windows optically polished
These cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a rigorous inspection process before leaving our factory. Fire-fused to ensure that cuvettes are robust and durable. Cuvette with a cylindrical chamber for polarimetric applications and a quartz-to-pyrex graded seal tube for adding additional structures to the top portion of the cell.
Delivery information: Includes PTFE stopper.