Cylindrical Polarimeter Cells with PTFE Stopper, FireflySci
Supplier: FIREFLYSCI, INC.
35G10 35G20 35G50 35G100 35UV10 35UV20 35UV50 35UV100 35IR10 35IR20 35IR50 35IR100
76015-960EA 586.24 CAD
76015-960 76015-962 76015-964 76015-966 76015-968 76015-970 76015-972 76015-974 76015-976 76015-978 76015-980 76015-982
Cylindrical Polarimeter Cells with PTFE Stopper, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality cylindrical polarimeter comes in several different materials and are a great value.
  • Comes in several different materials: (Optical Glass, UV quartz, IR quartz)
  • Lightpaths from 10 - 100 mm available
  • Two windows optically polished
Medium lightpath polarimeter cell with a large surface. These cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a rigorous inspection process before it leaves the factory. Fire-fused to ensure that cuvettes are robust and durable.
Delivery information: Includes PTFE stopper.