Stopcocks, High Vacuum, Straight Bore, Hollow Glass Plugs, Chemglass
Supplier: Chemglass
CG-471-01 CG-471-02
80074-306EA 334.77 CAD
80074-306 80074-308
Stopcocks, High Vacuum, Straight Bore, Hollow Glass Plugs, Chemglass
Stopcocks
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug.
Plug only part numbers are not supplied with retaining clip.
Delivery information: Supplied complete with the listed size CG-423 metal retaining clip.
Order Now
SPECIFICATIONS
Environmentally Preferable