Stopcocks, T-Bore, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-436-01 CG-436-02 CG-436-03
80074-210EA 298.68 CAD
80074-210 80074-212 80074-214
Stopcocks, T-Bore, PTFE Plugs, Chemglass
Stopcocks
A 1:5 taper PTFE stopcock with three stems sealed 90° apart. The PTFE plug has a 'T' shaped bore to connect any two, or all three stems.