Stopcocks, Straight Bore, 1-Arm Larger, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-428-01 CG-428-02 CG-428-03
80074-182EA 147.47 CAD
80074-182 80074-184 80074-186
Stopcocks, Straight Bore, 1-Arm Larger, PTFE Plugs, Chemglass
Stopcocks
Similar to the CG-424 series, but with one arm of the stopcock being of a larger diameter.
Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets. Available complete with 1:5 taper PTFE plug or glass shell only.
Ordering information: For replacement plugs, see the CG-424 series.