Material Rolls for Material Dispenser with Liner Remover, Brady
Supplier: Brady Worldwide
THT-500-403 THT-750-403 THT-1000-403 THT-1000-498 THT-1500-498 THT-500-499 THT-750-499 THT-1000-499 THT-1500-499 THT-1500-403 THT-2000-403 THT-500-498 THT-750-498
76319-624RL 85.04 CAD
76319-624 76319-626 76319-628 76319-630 76319-632 76319-634 76319-636 76319-638 76319-640 76319-714 76319-716 76319-718 76319-720
Material Rolls for Material Dispenser with Liner Remover, Brady
Labels
Thermal transfer white labels for the Brady® Material dispenser with liner remover.
  • Sticks to any clean, dry surface
  • Ideal for use in wire and cable identification, or temporary vial labeling
  • Recommended for indoor use
The B-402 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind. Ideal for use in laboratory settings, asset tracking, or general use. Easy removal saves time during re-labeling processes. Pressure-sensitive adhesive allows for quick and easy application.
Permanent or repositionable onceMore Product Information