Screw caps for Microcentrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
111671B 111671C 111671G 111671R 111671W 111671Y 111672B 111672C 111672G 111672R 111672W 111672Y
89501-434PK 348.25 CAD
89501-434 89501-436 89501-438 89501-442 89501-444 89501-446 89501-448 89501-450 89501-452 89501-454 89501-456 89501-458
Screw caps for Microcentrifuge Tubes, Globe Scientific
Closures Test/Sample Tube Closures
These screwcaps for microcentrifuge tubes are suitable for all Globe Scientific screw top microcentrifuge tubes.
  • Made of polypropylene
  • Nitrile o-ring
The caps are available in a variety of colors and are offered either with or without an o-ring.