Stopcocks, Straight Bore, Glass Plugs, 1-Arm Larger, Chemglass
Supplier: Chemglass
CG-402-03 CG-402-01 CG-402-02
80074-070EA 214.28 CAD
80074-070 80074-066 80074-068
Stopcocks, Straight Bore, Glass Plugs, 1-Arm Larger, Chemglass
Stopcocks
Similar to the CG-400 series, but with one arm of the stopcock being of a larger diameter.
Larger arm facilitates construction of stronger apparatus such as separatory funnels and burets.
Ordering information: Supplied complete with the listed size metal retaining clip CG-423. For replacement plugs, see the CG-400 listing.