Pipette Dropping Clear Glass Bottles
Supplier: Wards
CH0194A CH0194B CH0194C
470314-028EA 10.6 CAD
470314-028 470314-030 470314-032
Pipette Dropping Clear Glass Bottles
Bottles Dropping Bottles
Ideal for labs and classrooms in chemistry applications, these bottles are also great for storing, mixing, and dispensing essential oils.
  • Non-screw top allows for quick and easy withdrawal
These borosilicate glass bottles come equipped with an eye dropper pipet for clean dispensing of the stored contents. The clear glass allows for maximum visibility and display of contents.