Octadecamethylcyclononasiloxane ≥98.0% (by GC)

Supplier: TCI America
Warning

O0425-1ML O0425-5ML
TCO0425-1MLEA 991.78 CAD
TCO0425-1ML TCO0425-5ML
Octadecamethylcyclononasiloxane ≥98.0% (by GC)
Octadecamethylcyclononasiloxane
Formula: C₁₈H₅₄O₉Si₉
Boiling Pt: 188 °C (20 mmHg) (lit.)
Melting Pt: –27 °C (lit.)
MDL Number: MFCD30533789
CAS Number: 556-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8a273e339d34f1c94c3dfebe5e9c1c8c1ebc8f7&vid=c8a273e339d34f1c94c3dfebe5e9c1c8c1ebc8f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 98.0%
Specific Gravity (20/20) 0.9790 - 0.9830
Refractive Index n20/D 1.4050 - 1.4090


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR