LT400 Protective Eyewear, HexArmor
Supplier: HexArmor
11-22001-02 11-22002-02
76317-748CS 486.21 CAD
76317-748 76317-750
LT400 Protective Eyewear, HexArmor
Safety Glasses
The multipurpose LT400 series of protective eyewear features permanent anti-fog coatings, innovative lens technology, and a sleek, lightweight frame designed to fit multiple face shapes for a comfortable, slip-free fit.
  • Panoramic lens with integrated side protectors and brow guard for total coverage and protection
  • Rimless lens design with a wide field of vision
  • Open rail side arms for functional accessories
  • 100% UV protection and metal-free
  • Innovative lens geometry and side arms ensure a close fit for most face shapes
  • LTX® lens technology for premium optical clarity
Designed with every wearer in mind for a variety of tasks and applications, LT400 safety eyewear ensures optimum comfort, style, and performance while on the job. Available in multiple tints.
Certifications: Compliant with ANSI Z87.1.2015. High Impact Rated Z87+. CSA Z94.3. CE EN 166