Helix® 2087 Series Gloves, Hexarmor
Supplier: HexArmor
2087-XXL(11) 2087-XXS(5) 2087-XXXL(12)
76317-710CS 642.16 CAD
76317-710 76317-712 76317-714 76318-150 76318-152 76318-154 76318-156 76318-158
Helix® 2087 Series Gloves, Hexarmor
Gloves
Helix® 2087 series gloves feature a flexible, foam nitrile palm coating for superior grip and abrasion resistance. The 13-gauge, HPPE and fiberglass blend shell offering 360° cut protection and provides workers with ANSI/ISEA level A4 cut resistance.
  • Flexible foam nitrile palm coating provides superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. ProvidingMore Product Information