Helix® 2085 Series Gloves, Hexarmor
Supplier: Hexarmor
2085-M(8) 2085-XS(6) 2085-XXXL(12) 2085-L(9) 2085-XXL(11) 2085-XL(10) 2085-S(7) 2085-XXS(5)
76317-690CS 793.35 CAD
76317-690 76317-680 76317-694 76317-684 76317-692 76317-682 76317-688 76317-686
Helix® 2085 Series Gloves, Hexarmor
Gloves
Seamless, coated Helix® series 2085 gloves feature a 13-gauge, HPPE and fiberglass blend shell that offering 360° cut protection and provides ANSI/ISEA level A4 cut resistance. The flexible polyurethane palm coating provides superior grip and abrasion resistance in dry, wet, or oily situations while maintaining flexibility and durability.
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: EN 388. CE 4X43C. ANSI/ISEA Cut A4