VWR® Gel-Loading Pipette Tips
Supplier: VWR International
76321-562 76321-564 76321-566 76321-568 76321-814 76321-828
76321-562BG 115.07 CAD
76321-562 76321-564 76321-566 76321-568 76321-814 76321-828
VWR® Gel-Loading Pipette Tips
Pipette Tips
VWR® Gel-Loading Pipette Tips faciliate precision sample loading into gels.
  • Standard 96 tip format is multi-channel friendly
  • Certified free of RNase, DNase, and human DNA (lot-by-lot certificates available)
  • Sterile product is also pyrogen-free
Universal fit to accommodate most pipettes except Rainin LTS and Thermo Clip-Tip.
Ordering information: Choose from sterile or nonsterile formats, racked or bulk packaging, slim or MBP body styles.