Leak Resistant Urine Collection Containers with Patient I.D. Label, Globe Scientific
Supplier: Globe Scientific
6220 6238 6526 6528 6529 6236
76293-180CS 416.17 CAD
76293-180 76293-182 76293-184 76293-186 76293-188 76299-868
Leak Resistant Urine Collection Containers with Patient I.D. Label, Globe Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
These containers are the ultimate in leak-tight storage and transport of liquids.
  • Containers are produced from polypropylene (PP)
  • Caps are produced from polyethylene (PE)
  • Molded graduation marks
An expanded inner liner in the screwcap is designed to compress against the uppermost walls and top rim of the container, thereby providing an air-tight seal. They are ideal for use in hospital pneumatic tube systems.
Packaging: The 120 ml Tite-Rite containers with orange cap are supplied individually wrapped.