Helix® 2082 Series Gloves, Hexarmor
Supplier: HexArmor
2082-XXL(11) 2082-S(7)
76317-674CS 1170.26 CAD
76317-674 76318-200 76318-202 76318-204 76318-206
Helix® 2082 Series Gloves, Hexarmor
Gloves
Helix® series 2082 gloves feature a 13-gauge, flame-resistant aramid and wool blend shell offering 360° cut protection. This glove offers ANSI/ISEA A3 cut protection and is rated HRC 1 for arc flash protection. A flexible, FR-compliant neoprene/nitrile blend palm coating provides superior grip and abrasion resistance.
  • Flexible FR-compliant neoprene/nitrile blend palm coating
  • Hazard risk category HRC 1 arc flash protection
  • EN 407 certified with score of 412210
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
Helix® seriesMore Product Information