Helix® 2068 Series Gloves, Hexarmor
Supplier: HexArmor
2068X-XS(6) 2068X-XXL(11) 2068X-XXXL(12) 2068X-M(8)
76317-662CS 522.87 CAD
76317-662 76317-664 76317-666 76317-668 76317-672 76318-120 76318-122 76318-124
Helix® 2068 Series Gloves, Hexarmor
Gloves
Lightweight, durable, and flexible with a seamless construction, the Helix® series 2068 glove combines cut and puncture protection with abrasion resistance in a seamless coated glove. Featuring a 13-gauge HPPE and nylon blend shell offering 360° cut protection, the 2068 Series provides ANSI/ISEA level A3 cut resistance and a polyurethane palm coating for superior grip and abrasion resistance.
  • Flexible polyurethane palm coating provides superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
More Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Powdered/Powder-free Powder-free
Liner material HPPE/nylon HPPE/nylon
Finish Polyurethane Palm Polyurethane Palm
Cuff style Knit wrist Knit wrist