SLT-AH-Series Tables with Book Compartments, National Public Seating
Supplier: National Public Seating
SLT4-2460CB SLT4-2472CB SLT4-3060CB SLT5-2448C SLT5-2454C SLT5-2460C SLT5-2472C SLT5-3060C SLT5-2448CB SLT5-2454CB SLT5-2460CB SLT5-2472CB SLT5-3060CB SLT4-2448CC SLT4-2454CC SLT4-2460CC SLT4-2472CC SLT4-3060CC SLT4-2448CBC SLT4-2454CBC SLT4-2460CBC SLT4-2472CBC SLT4-3060CBC SLT5-2448CC SLT5-2454CC SLT5-2460CC SLT5-2472CC SLT5-3060CC SLT5-2448CBC SLT5-2454CBC SLT5-2460CBC SLT5-2472CBC SLT5-3060CBC SLT4-2448C SLT4-2454C SLT4-2460C SLT4-2472C SLT4-3060C SLT4-2448CB SLT4-2454CB
76276-546EA 2842.42 CAD
76276-546 76276-548 76276-550 76276-552 76276-554 76276-556 76276-558 76276-560 76276-562 76276-564 76276-566 76276-568 76276-570 76276-572 76276-574 76276-576 76276-578 76276-580 76276-582 76276-584 76276-586 76276-588 76276-590 76276-592 76276-594 76276-596 76276-598 76276-600 76276-602 76276-604 76276-606 76276-608 76276-610 76276-788 76276-790 76276-792 76276-794 76276-796 76276-798 76276-800
SLT-AH-Series Tables with Book Compartments, National Public Seating
Furniture Tables and Workbenches
These SLT-AH adjustable height science lab tables have perfect chemistry with any science classroom. These paragons of endurance are built with a choice of stylistic light oak woodgrain or striking textured black legs beneath a chem-res tabletop with an MDF Core. The work surface is of solid black, firmly secured, high-pressure laminate, resistant to water and 13 commonly used laboratory chemicals.
  • Adapts for ADA requirements
  • Patented mechanism allows table to rise effortlessly and automatically lock at your height setting requirement
  • Oversized matching floor glides are standard
  • Available with book compartments
  • 10-year warranty
Robust legs are 2.325" diameter and adjust smoothly in 0.75" increments from 29 to 41". Legs automatically lock at your height setting to conveniently accommodate all seating requirements. Easily assemble the SLT-AH thanks to pre-drilled pilot holes. Feet are outfitted with oversized glides, or optional casters. Optional book compartment models come with two storage compartments concealable with the included side-panel aprons.
Load capacity: 500 lbs.
Certifications: Meets MAS certified green environmental industrial standards.
Ordering information: Optional casters available.