Wood Frame Tables with Phenolic Top and Solid Wood Legs, National Public Seating
Supplier: National Public Seating
SLT1-2448P SLT1-2454P SLT1-2460P SLT2-3072PB SLT1-2472P SLT1-3060P SLT1-3072P SLT2-2448P SLT2-2454P SLT2-2460P SLT2-2472P SLT2-3060P SLT2-3072P SLT1-2448PB SLT1-2454PB SLT1-2460PB SLT1-3060PB SLT1-3072PB SLT2-2448PB SLT2-2454PB SLT2-2460PB SLT2-3060PB
76276-612EA 1886.41 CAD
76276-612 76276-614 76276-616 76276-618 76276-802 76276-804 76276-806 76276-808 76276-810 76276-812 76276-814 76276-816 76276-818 76276-820 76276-822 76276-824 76276-828 76276-830 76276-832 76276-834 76276-836 76276-840
Wood Frame Tables with Phenolic Top and Solid Wood Legs, National Public Seating
Furniture Tables and Workbenches
The brand-new phenolic science lab tables will have perfect chemistry with any science classroom. These paragons of endurance are built with strong ash frame and side panel aprons beneath a solid, black, phenolic resin tabletop with a matte finish.
  • Solid ash legs and apron
  • Rubber liquid protecting boots
  • Adjustable glides for accurate leveling
  • Black, 0.75" phenolic top are highly impervious to most chemicals, acids, corrosions, stains and moisture
  • Comparable to epoxy resin tops
  • 10-year warranty
The work surface is crack-resistant and can be modifiedMore Product Information