U-Demountable Cuvettes with Removable Windows, FireFlySci
Supplier: FIREFLYSCI, INC.
19UV0.05 19UV0.1 19UV0.2 19UV0.5 19UV1 19UV2 19IR0.05 19IR0.1 19IR0.2 19IR0.5 19IR1
76015-610EA 1310.97 CAD
76015-610 76015-612 76015-614 76015-616 76015-618 76015-620 76015-622 76015-624 76015-626 76015-628 76015-630
U-Demountable Cuvettes with Removable Windows, FireFlySci
Cuvettes Spectrophotometer Cuvettes
U-demountable cuvettes have two polished windows.
Ordering information: A20 cell mount not included.
Order Now
SPECIFICATIONS
Environmentally Preferable