Hei-VAP Core Rotary Evaporators, Heidolph®
Supplier: Heidolph NA, LLC
036040050 036040051 036040053 036040055 036040056 036040058 036040060 036040061 036040063 036040065 036040066 036040068
CA76320-914EA 9312.82 CAD
CA76320-914 CA76320-762 CA76320-766 CA76320-770 CA76320-772 CA76320-776 CA76320-930 CA76320-932 CA76320-802 CA76320-806 CA76320-808 CA76320-812
Hei-VAP Core Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed to access the basic parameters rotation speed and heating bath temperature at any time.
  • Clear digital display showing actual and set values
  • Accidental changing of the values can be prevented by the lock function
  • Standby button stops all functions
  • Standby function and residual heat warning at heating bath temperatures above 50 °C
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Overheat protection bath cut-off at 5 °C over set temperature via separate Pt1000
  • Secondary over temperature cut-off 250 °C
More Product Information