Macro 65 mm Tall Cuvettes with PTFE Cover, FireflySci

Supplier: FIREFLYSCI, INC.
508G10 508UV10
76016-410EA 282.79 CAD
76016-410 76016-412
Macro 65 mm Tall Cuvettes with PTFE Cover, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality macro cuvettes with spectrophotometer macro capacities and are a great value.

  • Fire–fused to ensure that cuvettes are robust and durable
  • Comes in several different materials including Optical Glass and UV quartz
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Includes PTFE cover
  • Extended height of 65 mm

FireflySci's Type 508 cuvette is a stretched-out version of the Type 1 cuvette. At 65mm tall, this cuvette is a good fit for many researchers with custom builds and machines. Highly accurate results, only need to test sample once. Firefly cuvettes are made with the best quality quartz and glass material available.

Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a vigorous inspection process before leaving the factory.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR