Replacement Components for 1:5 Taper PTFE Plugs, Chemglass
Supplier: Chemglass
CG-457-01 CG-457-02 CG-457-07 CG-457-08 CG-457-09 CG-457-10 CG-457-11 CG-457-12 CG-457-13 CG-457-14 CG-457-15 CG-457-16 CG-457-17 CG-457-18 CG-457-19 CG-457-20 CG-457-21
80063-076EA 39.67 CAD
80063-076 80063-078 80063-088 80063-090 80063-092 80063-094 80063-096 80063-098 80063-100 80063-102 80063-104 80063-106 80063-108 80063-110 80063-112 80063-114 80063-116
Replacement Components for 1:5 Taper PTFE Plugs, Chemglass
Seals Sealing Rings
Replacement components for 1:5 taper PTFE plugs. O-rings are viton, nuts are green, and washers are white.
Order Now
SPECIFICATIONS
Environmentally Preferable