Helix® 1095 Series Gloves, Hexarmor
Supplier: HexArmor
1095-L(9) 1095-M(8) 1095-S(7) 1095-XL(10) 1095-XS(6) 1095-XXL(11) 1095-XXS(5) 1095-XXXL(12)
76318-100CS 1409.39 CAD
76318-100 76318-102 76318-104 76318-106 76318-108 76318-110 76318-166 76318-168
Helix® 1095 Series Gloves, Hexarmor
Gloves
Helix® series 1095 nylon seamless knit gloves feature a back-of-hand IR-X® Exoskeleton™ impact protection system, a knit wrist to help prevent dirt and debris from entering the glove, and a silicone-free, flexible sandy nitrile palm coating. All elements combine to provide a superior grip and abrasion resistance on the job.
  • High-performance 15-gauge nylon blend shell
  • Flexible sandy nitrile palm coating provides superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Back-of-hand IR-X® Exoskeleton™ with high-
More Product Information