Helix® 1091 Series Gloves, Hexarmor
Supplier: HexArmor
1091-L(9) 1091-M(8) 1091-S(7) 1091-XL(10) 1091-XS(6) 1091-XXL(11)
76318-088CS 330.49 CAD
76318-088 76318-090 76318-092 76318-094 76318-096 76318-098
Helix® 1091 Series Gloves, Hexarmor
Gloves
Helix® Series 1091 nylon seamless knit glove features a knit wrist to help prevent dirt and debris from entering the glove. A silicone-free, flexible foam nitrile palm coating provides superior grip and abrasion resistance on the job.
  • High-performance 15-gauge nylon blend shell offering 360° cut protection
  • Flexible foam nitrile palm coating provides superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Silicone-free
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: EN 388 CE 4122X. ANSI/ISEA Cut A1.