Borosilicate Glass Burettes with Pinch Clips
Supplier: Wards
CH0234A CH0234B CH0234C CH0234D
470313-960EA 17.55 CAD
470313-960 470313-962 470313-964 470313-966
Borosilicate Glass Burettes with Pinch Clips
Burettes
Single bore burettes with detachable rubber tube and pinch clip.
  • Made of high quality 3.3 borosilicate glass
  • 0.10 ml sub divisions
  • Tolerance of ±0.20 ml
Certifications: DIN ISO 385 standard, Class B