Type 18M Micro Black Cuvettes with PTFE Cover, FireFlySci
Supplier: FIREFLYSCI, INC.
18MG5 18MG10 18MG20 18MG40 18MG50 18MUV5 18MUV10 18MUV20 18MUV40 18MUV50 18MIR5 18MIR10 18MIR20 18MIR40 18MIR50
76015-580EA 657.01 CAD
76015-580 76015-582 76015-584 76015-586 76015-588 76015-590 76015-592 76015-594 76015-596 76015-598 76015-600 76015-602 76015-604 76015-606 76015-608
Type 18M Micro Black Cuvettes with PTFE Cover, FireFlySci
Cuvettes Spectrophotometer Cuvettes
The FireflySci Type 18M Micro Cuvette is a modified version of the Type 18 but with black walls for blocking out stray light.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in several different materials: (Optical Glass, UV Quartz and IR Quartz)
  • Lightpaths from 5 - 50 mm available
  • Two windows optically polished
  • Contains black walls to block out stray light
  • Base thickness - 3 mm
Firefly cuvettes are made with the best quality quartz and glass material available. Hogh transmission values.
Each cell goes through a rigorous inspection process before leaving the factory.