Disodium hydrogen phosphate ≥99% (by titrimetric analysis), Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

567547-1KG 567547-250GM
CA80503-410EA 394.86 CAD
CA80503-410 CA80503-412
Disodium hydrogen phosphate ≥99% (by titrimetric analysis), Molecular Biology Grade, Calbiochem®
di-Sodium hydrogen phosphate

Used in conjunction with sodium phosphate, monobasic in the preparation of biological buffers.

Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805

Order Now

Specification Test Results

Purity ≥99% (Titration)
DNases, proteases, RNases none detected
Absorbance (260 nm and 280 nm) ≤0.01
Heavy metals ≤0.001% (as Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR