Glass Microscope Slides, Azer Scientific
Supplier: Azer Scientific
2752510A 2752512A 2752511
76268-418CS 569.5 CAD
76268-418 76076-736 76076-734
Glass Microscope Slides, Azer Scientific
Slides Microscope Slides
General-purpose microscope slides offer quality and value for any laboratory.
  • Extra white soda-lime glass with 90 ° corners
  • Manufactured in Germany
Choose any one of these general-purpose microscope slides for clinical, environmental, industrial, or other routine microscopy work. Extra white soda-lime glass will not interfere with fluorescent staining procedures. Uniform 1.0 mm thickness makes these sturdy enough for field work.
Packaging: Slides are packaged 72 per box, 20 boxes per case.