Ward's® White's Tree Frog
Supplier: Avantor
470312-390PK 229 CAD
470312-390 470312-402
Ward's® White's Tree Frog
Specimens Live Specimens Animals - Living Material
Your frog is shipped in a plastic container with damp sphagnum moss as cushioning. It should be placed in a prepared habitat shortly after arrival.
  • Litoria caerulea
Ordering information: These products contain living or perishable materials. Visit https://wardsci.com/cms/teacher_live_materials for care instructions.
Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.