Helix® 1073 Series Gloves, Hexarmor
Supplier: HexArmor
1073-L(9) 1073-M(8) 1073-S(7) 1073-XL(10) 1073-XS(6) 1073-XXL(11) 1073-XXS(5) 1073-XXXL(12)
76318-072CS 1065.46 CAD
76318-072 76318-074 76318-076 76318-078 76318-080 76318-082 76318-084 76318-086
Helix® 1073 Series Gloves, Hexarmor
Gloves
Helix® 1073 series gloves protect but feel like a second skin, with a smooth finish microfoam nitrile palm coating. Made with a 15-gauge HPPE and fiberglass blended shell offering 360° cut protection, workers will experience excellent comfort and dexterity, allowing hands to move freely and safely on the job.
  • Microfoam nitrile palm coating provides superior grip and abrasion resistance
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Reinforced thumb crotch area for increased durability
  • Seamless construction for enhanced comfort and breathability
  • Silicone-free
Helix® series gloves offer a wide variety of grip options to provide users with the appropriate grip for the task at hand. Providing coverage for any working condition, HexArmor is constantly researching and testing new formulas and application methods to maximize grip and function, helping workers redefine safety at their worksite.
Certifications: EN 388 CE 4544C. ANSI/ISEA Cut A3.