Bottom Raised Micro Cuvettes with PTFE Cover, FireFlySci
Supplier: FIREFLYSCI, INC.
18BUV5 18BUV10 18BUV20 18BUV40 18BUV50 18BIR5 18BIR10 18BIR20 18BIR40 18BIR50
76015-476EA 585.04 CAD
76015-476 76015-478 76015-480 76015-482 76015-484 76015-486 76015-488 76015-490 76015-492 76015-494
Bottom Raised Micro Cuvettes with PTFE Cover, FireFlySci
Cuvettes Spectrophotometer Cuvettes
Bottom Raised Micro Cuvettes have two polished windows.
  • Base thickness is 9 mm
  • No black masking