Ball Mills, PTA, Nidec Shimpo
Supplier: Nidec Shimpo America
PTA-01 PTA-02
76307-536EA 4103.69 CAD
76307-536 76307-538
Ball Mills, PTA, Nidec Shimpo
Mills
Compact design that is ideal for small-scale production and is constructed using powder-coated steel for long-lasting durability.
  • Easy adjustments can be made to the sturdy roller bars to accommodate jars from 1 to 10 l
  • Comes standard with a breaker switch
Heavy-duty ball milling machines enable very precise milling, in porcelain jars, even for relatively tough materials, wet or dry.